EMAIL THIS PAGE TO A FRIEND
Oncology reports
CXCL7 promotes proliferation and invasion of cholangiocarcinoma cells.
PMID 27959418
Abstract
CXCL7 is an important chemoattractant cytokine, which signals through binding to its receptor CXCR2. Recent studies have demonstrated that the CXCL7/CXCR2 signaling plays a promoting role in several common malignancies, including lung, renal, colon, and breast cancer. However, the regulatory role of CXCL7, in cholangiocarcinoma, as well as the underlying mechanism, has not been previously reported. Herein, we found more positive expression of CXCL7 in cholangiocarcinoma tissues compared to adjacent non-tumor tissues. High CXCL7 expression was significantly correlated with poor differentiation, lymph node metastasis, vascular invasion and advanced clinical stage, but was not associated with age, gender, or tumor size. Besides, the expression of CXCL7 was significantly associated with the Ki67 expression, but not associated with CA199, AFP, or P53 expression in cholangiocarcinoma. Moreover, the overall survival of cholangiocarcinoma patients with high CXCL7 expression was significantly shorter than those with low CXCL7 expression. Inxa0vitro study indicated that CXCL7 and CXCR2 were also positively expressed in several common cholangiocarcinoma cell lines, including HuCCT1, HuH28, QBC939, EGI-1, OZ and WITT. SiRNA-induced inhibition of CXCL7 significantly reduced the proliferation and invasion of QBC939 cells. On the contrary, overexpression of CXCL7 markedly promoted these malignant phenotypes of QBC939 cells. Of note, the conditioned medium of CXCL7-overexpresing human hepatic stellate cells could also promote the proliferation and invasion of QBC939 cells, suggesting that CXCL7 may also play an oncogenic role in cholangiocarcinoma in a paracrine-dependent manner, not only in an autocrine-dependent manner. Molecular assay data suggested that the AKT signaling pathway was involved in the CXCL7-mediated malignant phenotypes of QBC939 cells. In summary, our study suggests that CXCL7 plays a promoting role in regulating the growth and metastasis of cholangiocarcinoma.
Related Materials