Oncology reports
17β-estradiol-induced growth of triple-negative breast cancer cells is prevented by the reduction of GPER expression after treatment with gefitinib.
PMID 27959426
Triple-negative breast cancers (TNBCs) are neither susceptible to endocrine therapy due to a lack of estrogen receptorxa0α expression nor trastuzumab. TNBCs frequently overexpress epidermal growth factor receptor (EGFR) and membrane bound estrogen receptor, GPER. To a certain extent the growth of TNBCs is stimulated by 17β-estradiol via GPER. We analyzed whether inhibition of EGFR by gefitinib reduces the expression of GPER and subsequent signal transduction in TNBC cells. Dependence of proliferation on 17β-estradiol was determined using Alamar Blue assay. Expression of GPR30 and activation of c-src, EGFR and cAMP-responsive element binding (CREB) protein by 17β-estradiol was analyzed by western blotting. Expression of c-fos, cyclinxa0D1 and aromatase was determined using RT-PCR. Gefitinib reduced GPER expression concentration‑xa0and time‑dependently. In HCC70 cells, GPER expression was reduced to 15±11% (p<0.05) after treatment with 200xa0nM gefitinib for four days, and in HCC1806 cells GPER expression was reduced to 39±5% (p<0.01) of the control. 17β-estradiol significantly increased the percentage of HCC1806 cells within 7xa0days to 145±29% of the control (HCC70, 110±8%). This increase in cell growth was completely prevented in both TNBC cell lines after GPR30 expression was downregulated by treatment with 200xa0nM gefitinib. In HCC1806 cells, activation of c-src was increased by 17β-estradiol to 350±50% (p<0.01), and gefitinib reduced src activation to 110%. Similar results were obtained in the HCC70 cells. Phosphorylation of EGFR increased to 240±40% (p<0.05) in the HCC1806 cells treated with 17β-estradiol (HCC70,xa0147±25%). Gefitinib completely prevented this activation. Phosphorylation of CREB and induction of c-fos, cyclin D1 and aromatase expression by 17β-estradiol were all prevented by gefitinib. These experiments conclusively show that reduction of GPER expression is a promising therapeutic approach for TNBC.