EMAIL THIS PAGE TO A FRIEND
The New England journal of medicine
Prevention of Bleeding in Patients with Atrial Fibrillation Undergoing PCI.
PMID 27959713
Abstract
In patients with atrial fibrillation undergoing percutaneous coronary intervention (PCI) with placement of stents, standard anticoagulation with a vitamin K antagonist plus dual antiplatelet therapy (DAPT) with a P2Y We randomly assigned 2124 participants with nonvalvular atrial fibrillation who had undergone PCI with stenting to receive, in a 1:1:1 ratio, low-dose rivaroxaban (15 mg once daily) plus a P2Y The rates of clinically significant bleeding were lower in the two groups receiving rivaroxaban than in the group receiving standard therapy (16.8% in group 1, 18.0% in group 2, and 26.7% in group 3; hazard ratio for group 1 vs. group 3, 0.59; 95% confidence interval [CI], 0.47 to 0.76; P<0.001; hazard ratio for group 2 vs. group 3, 0.63; 95% CI, 0.50 to 0.80; P<0.001). The rates of death from cardiovascular causes, myocardial infarction, or stroke were similar in the three groups (Kaplan-Meier estimates, 6.5% in group 1, 5.6% in group 2, and 6.0% in group 3; P values for all comparisons were nonsignificant). In participants with atrial fibrillation undergoing PCI with placement of stents, the administration of either low-dose rivaroxaban plus a P2Y