Journal of medical microbiology
Cell cycle arrest and apoptosis induced by enteroaggregative Escherichia coli in cultured human intestinal epithelial cells.
PMID 27959791
Enteroaggregative Escherichia coli (EAEC) is an emerging enteric pathogen causing diarrhoeal diseases in multiple epidemiological and clinical settings. However, understanding of the pathogenesis of the disease caused by this organism is still suboptimal. Studies have indicated that enteric bacteria induced cell cycle arrest and apoptosis in host intestinal epithelial cells might play a vital role in the pathogenesis caused by these organisms. In this study an attempt was made to assess EAEC-induced apoptosis and cell cycle modulation in human intestinal epithelial cell lines. INT-407 and HCT-15 cells were infected with EAEC-T8 (clinical isolate) as well as plasmid cured variant of EAEC-T8 (EAEC-pT8). Propidium iodide staining was done to select the time of infection and the incubation period of the infected culture. Apoptosis was further assessed in EAEC infected both the cell lines by annexin-V-FLUOS & propidium iodide, cell death detection ELISA, DNA strand breaks and microscopic analysis. Further, the DNA content of the EAEC-infected cells at different phases of cell cycle was also monitored. We have found that EAEC could induce apoptosis in human small intestinal as well as colonic epithelial cell lines, which was assessed by the expression of phosphatidylserine on host cell surface, internucleosomal cleavage of host cell DNA and microscopic analysis of the characteristic apoptotic features of these cells. EAEC was also found to arrest cells at S phase and G2-M phase of the cell cycle. EAEC-T8 could induce maximum apoptosis and cell cycle modulation in both small intestinal and colonic epithelial cells. Further, we have observed that the plasmid of this organism had maximum contribution to these processes. The outcome of this study has undoubtedly led to a better understanding of the basic mechanism of pathogenesis caused by EAEC.