EMAIL THIS PAGE TO A FRIEND
Journal of water and health
Antibacterial and antiviral effectiveness of two household water treatment devices that use monobrominated hydantoinylated polystyrene.
PMID 27959873
Abstract
Many different household water treatment (HWT) methods have been researched and promoted to mitigate the serious burden of diarrheal disease in developing countries. However, HWT methods using bromine have not been extensively evaluated. Two gravity-fed HWT devices (AquaSure™ and Waterbird™) were used to test the antimicrobial effectiveness of HaloPure