EMAIL THIS PAGE TO A FRIEND
Environmental toxicology and pharmacology
Tebufenozide induces G1/S cell cycle arrest and apoptosis in human cells.
PMID 27960113
Abstract
Tebufenozide is a non-steroidal insect growth regulator and is extensively used to control pests, although it is considered to be safe for mammals and environmentally friendly. However, previous studies have found that tebufenozide is cytotoxic to man, although the exact mechanism remains elusive. This study will investigate the apoptotic molecular mechanisms which result from tebufenozide-induced DNA damage in HeLa cells. Our results demonstrate that tebufenozide could trigger arrest in G1/S phase related to a downregulation of cyclin E and cyclin-dependent kinase (CDK) 2 protein. In addition, Western blotting showed apoptosis was associated with the upregulation of p53, Bax and cleaved-PARP, as well as downregulation of Bcl-2 and PARP. Tebufenozide also regulated changes in mitochondrial permeability and reduced mitochondrial number and intracellular ATP production. Briefly, these results suggest that tebufenozide- induces cell cycle arrest and apoptosis through activating p53 protein in a Bax- and Bcl-2-triggered mitochondrial pathway. This work provides some scientific context for the safe use of tebufenozide in agriculture.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
31652
Tebufenozide, PESTANAL®, analytical standard
C22H28N2O2