EMAIL THIS PAGE TO A FRIEND
The Japanese journal of psychiatry and neurology
Concentrations of beta-phenylethylamine in plasma and plateletes of schizophrenics.
PMID 2796027
Abstract
The plasma and platelet PEA levels of 20 normal subjects and 17 schizophrenic patients were investigated using a high-performance liquid chromatography. In the normals the mean plasma and platelet levels of PEA were 4.9 +/- 1.9 ng/ml and 1.78 +/- 1.01 ng/mg protein, respectively, while in the schizophrenics, those were 12.1 +/- 7.9 ng/ml and 0.77 +/- 0.5 ng/mg protein, respectively. The plasma PEA levels of the schizophrenics were significantly higher than those of the normals, and the platelet PEA levels of the schizophrenics were lower than those of the normals.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
407267
Phenethylamine, purified by redistillation, ≥99.5%
C8H11N