EMAIL THIS PAGE TO A FRIEND
European journal of vascular and endovascular surgery : the official journal of the European Society for Vascular Surgery
Pre-operative Simulation of the Appropriate C-arm Position Using Computed Tomography Post-processing Software Reduces Radiation and Contrast Medium Exposure During EVAR Procedures.
PMID 27965020
Abstract
The aim was to evaluate the feasibility and efficacy of a new method for pre-operative calculation of an appropriate C-arm position for iliac bifurcation visualisation during endovascular aortic repair (EVAR) procedures by using three dimensional computed tomography angiography (CTA) post-processing software. Post-processing software was used to simulate C-arm angulations in two dimensions (oblique, cranial/caudal) for appropriate visualisation of distal landing zones at the iliac bifurcation during EVAR. Retrospectively, 27 consecutive EVAR patients (25 men, mean ± SD age 73 ± 7 years) were identified; one group of patients (NEW; n = 12 [23 iliac bifurcations]) was compared after implementation of the new method with a group of patients who received a historic method (OLD; n = 15 [23 iliac bifurcations]), treated with EVAR before the method was applied. In the OLD group, a median of 2.0 (interquartile range [IQR] 1-3) digital subtraction angiography runs were needed per iliac bifurcation versus 1.0 (IQR 1-1) runs in the NEW group (p = .007). The median dose area products per iliac bifurcation were 11951 mGy*cm Pre-operative simulation of the appropriate C-arm angulation in two dimensions using dedicated computed tomography angiography post-processing software is feasible and significantly reduces radiation and contrast medium exposure.