EMAIL THIS PAGE TO A FRIEND
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
Dietary intake of phosphorus flame retardants (PFRs) using Swedish food market basket estimations.
PMID 27965106
Abstract
The occurrence of eight phosphorus flame retardants (PFRs) was investigated in 53 composite food samples from 12 food categories, collected in 2015 for a Swedish food market basket study. 2-ethylhexyl diphenyl phosphate (EHDPHP), detected in most food categories, had the highest median concentrations (9 ng/g ww, pastries). It was followed by triphenyl phosphate (TPHP) (2.6 ng/g ww, fats/oils), tris(1,3-dichloro-2-propyl) phosphate (TDCIPP) (1.0 ng/g ww, fats/oils), tris(2-chloroethyl) phosphate (TCEP) (1.0 ng/g ww, fats/oils), and tris(1-chloro-2-propyl) phosphate (TCIPP) (0.80 ng/g ww, pastries). Tris(2-ethylhexyl) phosphate (TEHP), tri-n-butyl phosphate (TNBP), and tris(2-butoxyethyl) phosphate (TBOEP) were not detected in the analyzed food samples. The major contributor to the total dietary intake was EHDPHP (57%), and the food categories which contributed the most to the total intake of PFRs were processed food, such as cereals (26%), pastries (10%), sugar/sweets (11%), and beverages (17%). The daily per capita intake of PFRs (TCEP, TPHP, EHDPHP, TDCIPP, TCIPP) from food ranged from 406 to 3266 ng/day (or 6-49 ng/kg bw/day), lower than the health-based reference doses. This is the first study reporting PFR intakes from other food categories than fish (here accounting for 3%). Our results suggest that the estimated human dietary exposure to PFRs may be equally important to the ingestion of dust.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M47949
2-Methylheptane, 98%
C8H18
35586
PCB No 1, analytical standard
C12H9Cl