EMAIL THIS PAGE TO A FRIEND
Scientific reports
New Immunosuppressive Sphingoid Base and Ceramide Analogues in Wild Cordyceps.
PMID 27966660
Abstract
A comprehensive identification of sphingoid bases and ceramides in wild Cordyceps was performed by integrating a sequential chromatographic enrichment procedure and an UHPLC-ultrahigh definition-Q-TOF-MS based sphingolipidomic approach. A total of 43 sphingoid bases and 303 ceramides were identified from wild Cordyceps, including 12 new sphingoid base analogues and 159 new ceramide analogues based on high-resolution MS and MS/MS data, isotope distribution, matching with the comprehensive personal sphingolipid database, confirmation by sphingolipid standards and chromatographic retention time rule. The immunosuppressive bioassay results demonstrated that Cordyceps sphingoid base fraction exhibits more potent immunosuppressive activity than ceramide fraction, elucidating the immunosuppressive ingredients of wild Cordyceps. This study represented the most comprehensive identification of sphingoid bases and ceramides from a natural source. The findings of this study provided an insight into therapeutic application of wild Cordyceps.