Reproduction in domestic animals = Zuchthygiene
Oxytocin receptors in dioestrous and anoestrous canine uteri.
PMID 27966823
The aim of the study was to localize oxytocin receptors (OTR) and measure mRNA expression of OTR in the canine uterus with and without the influence of progesterone. Uterine samples were taken from nine anoestrous and eight dioestrous bitches during ovariohysterectomy. Histological changes were evaluated in haematoxylin and eosin (HE)-stained samples. Purified polyclonal antibody for OTR was used in immunohistochemistry to localize receptors in uterine layers. Relative mRNA concentration of OTR was evaluated with real-time PCR from full-thickness uterine samples taken from the middle horn and the body. Myometrial smooth muscle cells, endometrial luminal epithelium (LE) and deep and superficial glandular epithelium were positively stained for oxytocin receptors in non-pregnant animals. No significant difference in staining intensity was detected between uterine middle horn and body. However, the staining intensity of LE was significantly higher in dioestrous than in anoestrous uteri (pxa0