EMAIL THIS PAGE TO A FRIEND
Water science and technology : a journal of the International Association on Water Pollution Research
Fabrication of sulfated nanofilter membrane based on carboxymethyl cellulose.
PMID 27973365
Abstract
The aim of this study is to prepare sulfated carboxymethyl cellulose (SCMC) nanofilter membrane using sulfur trioxide pyridine complex (SO
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A4562
5-(N,N-Dimethyl)amiloride hydrochloride
C8H12ClN7O · HCl