EMAIL THIS PAGE TO A FRIEND
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology
An approach to utilize the artificial high power LED UV-A radiation in photoreactors for the degradation of methylene blue.
PMID 27982156
Abstract
Utilization of UV LED light is trending in the development of photoreactors for pollutant treatment. In this study, two different geometries were studied in the degradation of methylenebBlue (MB) using high power UVA LED as a source of light. The dosage, initial concentration, electric power, and H
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
38800
Malachite Green chloride, analytical standard
C23H25ClN2