EMAIL THIS PAGE TO A FRIEND
Analytical chemistry
Constant Ion Loss Method for the Untargeted Detection of Bis-sulfate Metabolites.
PMID 27982580
Abstract
The untargeted detection of phase II metabolites is a key issue for the study of drug metabolism in biological systems. Sensitive and selective mass spectrometric (MS) techniques coupled to ultrahigh performance liquid chromatographic (UHPLC) systems are the most effective for this purpose. In this study, we evaluate different MS approaches with a triple quadrupole instrument for the untargeted detection of bis-sulfate metabolites. Bis-sulfates of 23 steroid metabolites were synthesized and their MS behavior was comprehensively studied. Bis-sulfates ionized preferentially as the dianion ([M - 2H]
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0941 Tibolone, ≥98% (HPLC)
C21H28O2