EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
Design and Synthesis of a Pan-Janus Kinase Inhibitor Clinical Candidate (PF-06263276) Suitable for Inhaled and Topical Delivery for the Treatment of Inflammatory Diseases of the Lungs and Skin.
PMID 27983835
Abstract
By use of a structure-based computational method for identification of structurally novel Janus kinase (JAK) inhibitors predicted to bind beyond the ATP binding site, a potent series of indazoles was identified as selective pan-JAK inhibitors with a type 1.5 binding mode. Optimization of the series for potency and increased duration of action commensurate with inhaled or topical delivery resulted in potent pan-JAK inhibitor 2 (PF-06263276), which was advanced into clinical studies.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
PZ0324
PF-06263276, ≥98% (HPLC)
C31H31FN8O2