EMAIL THIS PAGE TO A FRIEND
Progress in clinical and biological research
Inhibition of actin-myosin-ATP interaction of smooth muscle native actomyosin is not closely associated with inhibition of its myosin light chain phosphorylation: studies with derivatives of antibiotic SF-2370.
PMID 2798505