EMAIL THIS PAGE TO A FRIEND
International journal of biological macromolecules
Unraveling the interaction of hemoglobin with a biocompatible and cleavable oxy-diester-functionalized gemini surfactant.
PMID 27986633
Abstract
Surfactant-protein mixtures have attracted considerable research interest in recent years at the interface of chemical biology and medicinal chemistry. Herein, the interaction between a green gemini surfactant (C
Related Materials