EMAIL THIS PAGE TO A FRIEND
Carbohydrate polymers
Role of nanostructured aggregation of chitosan derivatives on [5-methionine]enkephalin affinity.
PMID 27987934
Abstract
Affinities of quaternary ammonium-chitosan conjugates, their thiolated derivatives and corresponding nanostructured aggregates towards the hydrophilic drug [5-methionine]enkephalin were compared by Nuclear Magnetic Resonance (NMR) spectroscopic methods based on proton selective relaxation rate measurements. Nanoaggregates showed enhanced drug affinity in comparison with corresponding polymers, especially in the case of thiolated systems.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M6638
[Met5]Enkephalin acetate salt hydrate, ≥95.0% (HPLC), powder
C27H35N5O7S · xC2H4O2 · yH2O