EMAIL THIS PAGE TO A FRIEND
Carbohydrate polymers
Chitosan-based core-shell nanomaterials for pH-triggered release of anticancer drug and near-infrared bioimaging.
PMID 27987935
Abstract
As a naturally-abundant biopolymer, chitosan (CS) exhibit pH-sensitive structural transformation within a narrow pH range. Integrating hydrophobic groups to CS molecules gives modified CS polymers with more adjustable pH responsiveness. In this paper, near-infrared (NIR) photoluminescent Ag
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
O1008
Oleic acid, ≥99% (GC)
C18H34O2