EMAIL THIS PAGE TO A FRIEND
Carbohydrate polymers
Structural elucidation of three antioxidative polysaccharides from Tricholoma lobayense.
PMID 27987953
Abstract
Tricholoma lobayense is a nutritious mushroom with great health benefits. Three polysaccharides with purity higher than 99% were successfully extracted from Tricholoma lobayense. The molecular weights of TLH-1, TLH-2 and TLH-3 were determined to be 8.43×10
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W373011 L-Rhamnose, natural sourced, 99%, FG
C6H12O5H2O