EMAIL THIS PAGE TO A FRIEND
Carbohydrate polymers
Extraction, characterization and biological activity of a (1,3)(1,6)-β-d-glucan from the pathogenic oomycete Pythium insidiosum.
PMID 27987983
Abstract
Pythiosis is a life-threatening infectious disease caused by the pathogenic oomycete Pythium insidiosum. This study is the first to evaluate the P. insidiosum glucan content and its biological activities. The enzymatic quantification of the glucans in P. insidiosum mycelia showed that the β-glucan content was 18.99%±3.59. The cell wall polysaccharide extract consisted of ∼81.7% carbohydrates (exclusively glucose) and ∼18.3% residual amino acids and peptides. The results from monosaccharide composition, methylation and 1D/2D NMR spectroscopy analyses indicated the presence of a highly branched (1,3)(1,6)-β-d-glucan, with (1,6)-β-d-glucopyranosil side-branching unit on average every 1-2 repeat units. In vitro, the β-d-glucan extract could significantly promote spleen lymphocyte proliferation in human, equine and mouse cell cultures. BALB/c mice that were subcutaneously pre-immunized with three doses of 0.5, 2.5 and 5.0mg of β-glucan/mouse, showed a significant increase in IL-2, IL-6, IL-10, TNF-α and IL-17A production compared to non-immunized mice. These results suggested that β-d-glucan extract induces significant and specific Th17 cellular immune response and provided the theoretical basis for further experiments.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
693839
6-Phosphonohexanoic acid, 97%
C6H13O5P