EMAIL THIS PAGE TO A FRIEND
Carbohydrate polymers
Structural elucidation and immunostimulatory activity of polysaccharide isolated by subcritical water extraction from Cordyceps militaris.
PMID 27987993
Abstract
Water-soluble polysaccharides were obtained from Cordyceps militaris (C. militaris) (CMP) by subcritical water extraction (SWE). Two polysaccharides fractions, CMP-W1 and CMP-S1, were isolated from CMP using DEAE-52 cellulose and Sephadex G-150 column chromatography. The structural characteristics of CMP-W1 and CMP-S1 were investigated. The results showed that the molecular weight of CMP-W1 and CMP-S1 are 3.66×105Da and 4.60×105Da, respectively, and both of them were heteropolysaccharides composed of d-mannose, d-glucose, d-galactose with the molar ratios of 2.84:1:1.29 and 2.05:1:1.09, respectively. FT-IR spectra analysis suggested that CMP-W1 and CMP-S1 belonged to pyranose form sugar and protein free. For immunostimulatory activity assay in vitro, CMP-W1 and CMP-S1 significantly promoted lymphatic spleen cell proliferation of mice. Therefore, the polysaccharides obtained from C. militaris by SWE can be used as potential natural immunostimulant in functional foods or medicine.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W373011 L-Rhamnose, natural sourced, 99%, FG
C6H12O5H2O