EMAIL THIS PAGE TO A FRIEND
Carbohydrate polymers
Co-loading and intestine-specific delivery of multiple antioxidants in pH-responsive microspheres based on TEMPO-oxidized polysaccharides.
PMID 27988000
Abstract
In this study, pH-responsive microspheres loaded with multiple antioxidants were developed for intestine-specific delivery and exhibited synergistic activity. They consist of chitosan (CS)-coated microspheres made of TEMPO-oxidized Konjac glucomannan (OKGM) polymers, of which the carboxyl (COO
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
22040
β-Carotene, ≥97.0% (UV)
C40H56