EMAIL THIS PAGE TO A FRIEND
Carbohydrate polymers
Novel cellulose-based amorphous solid dispersions enhance quercetin solution concentrations in vitro.
PMID 27988001
Abstract
Quercetin (Q) is a bioactive flavonol with potential to benefit human health. However, Q bioavailability is relatively low, due to its poor aqueous solubility and extensive phase-II metabolism. Strategies to increase solution concentrations in the small intestinal lumen have the potential to substantially increase Q bioavailability, and by extension, efficacy. We aimed to achieve this by incorporating Q into amorphous solid dispersions (ASDs) with cellulose derivatives. Q was dispersed in matrices of cellulose esters including 6-carboxycellulose acetate butyrate (CCAB), hydroxypropylmethylcellulose acetate succinate (HPMCAS) and cellulose acetate suberate (CASub) to afford ASDs that provided stability against crystallization, and pH-triggered release. Blends of CASub and CCAB with the hydrophilic polyvinylpyrrolidone (PVP) further enhanced dissolution. The ASD 10% Q:20% PVP:70% CASub most significantly enhanced Q solution concentration under intestinal pH conditions, increasing area under the concentration/time curve (AUC) 18-fold compared to Q alone. This novel ASD method promises to enhance Q bioavailability in vivo.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Q4951
Quercetin, ≥95% (HPLC), solid
C15H10O7
S4701
Suplatast tosylate, ≥98% (HPLC)
C16H26SNO4 · C7H7SO3