EMAIL THIS PAGE TO A FRIEND
Regulatory toxicology and pharmacology : RTP
Delamanid is not metabolized by Salmonella or human nitroreductases: A possible mechanism for the lack of mutagenicity.
PMID 27988333
Abstract
Nitro-containing compounds such as nitrofuran and nitroimidazole are drugs used for the treatment of infectious diseases. However, many of these nitro-containing drugs are positive in the bacterial reverse mutation assay (Ames test). The recently approved anti-multidrug-resistant tuberculosis (MDR-TB) drug, delamanid (Deltyba™; OPC-67683), a derivative of 4-nitroimidazole, was negative for mutagenicity in the Ames assay. In Salmonella typhimurium, mutagenicity of nitro compounds has been closely associated with the ability of nitroreductase to metabolize (degradation)these compounds. To explore the lack of mutagenicity for delamanid, we examined the initial metabolic rate and mutagenic-specific activity of a series of nitro compounds in S. typhimurium TA100. The order of maximum mutagenic-activity was nitrofuran > 2-nitroimidazole > 5-nitroimidazole ≥ 4-nitroimidazole, which is very similar to the order of initial metabolic rate, i.e., the Pearson's correlation coefficient (r = 0.85) showed a correlation between metabolic rate and mutagenic-activity. No metabolism of delamanid was detected even after 60 h of treatment. In addition, delamanid was not reduced by two human nitroreductases. These facts may explain the absence of genotoxicity of delamanid in both in vitro and in vivo tests.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
195650
2-Nitroimidazole, 98%
C3H3N3O2