EMAIL THIS PAGE TO A FRIEND
Colloids and surfaces. B, Biointerfaces
α-Tocopheryl linolenate solid lipid nanoparticles for the encapsulation, protection, and release of the omega-3 polyunsaturated fatty acid: in vitro anti-melanoma activity evaluation.
PMID 27988473
Abstract
The main target of this study was the preparation, characterization and antioxidant activity evaluation of α-tocopheryl linolenate based solid lipid nanoparticles (SLNs-TL), able to incorporate omega-3 α-linolenic acid, useful for the treatment of melanoma, a type of skin cancer. In particular, α-linolenic acid was successfully derivatized with α-tocopherol and the obtained compound was characterized by Fourier transform infrared (FT-IR) and by
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
102245
2-(Dimethylamino)pyridine, 97%
C7H10N2