EMAIL THIS PAGE TO A FRIEND
International journal of biological macromolecules
Improvement of chitinase Pachi with nematicidal activities by random mutagenesis.
PMID 27989482
Abstract
Chitinase, an enzyme that can degrade the main compositions of insect intestine and cuticle, has been used in the bio-control field. Our previous work has reported the chitinase Pachi with nematicidal activity (Caenorhabditis elegans). In the present study, to improve the chitinolytic and nematicidal activities of Pachi, a random mutant library was constructed by error-prone PCR and screened by bacteriophage T7-based high-throughput screening system. One mutant, Pachi
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I6758
IPTG, ≥99% (TLC), ≤0.1% Dioxane
C9H18O5S