EMAIL THIS PAGE TO A FRIEND
Laboratory investigation; a journal of technical methods and pathology
Hepatic stimulator substance inhibits calcium overflow through the mitochondria-associated membrane compartment during nonalcoholic steatohepatitis.
PMID 27991906
Abstract
Nonalcoholic fatty liver disease is considered a disorder of the endoplasmic reticulum (ER) and mitochondria. Recent studies have shown that the ER and mitochondrial membranes overlap by 15-20%, a region referred to as the 'mitochondria-associated ER membrane' (MAM). Some proteins, including sarco/ER calcium ATPase (SERCA), are located in the MAM and have an important role in Ca
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
77540
o-Phenetidine, ≥97.0% (GC)
C8H11NO