EMAIL THIS PAGE TO A FRIEND
EMBO reports
Glioma-derived cancer stem cells are hypersensitive to proteasomal inhibition.
PMID 27993939
Abstract
Although proteasome inhibitors (PIs) are used as anticancer drugs to treat various cancers, their relative therapeutic efficacy on stem cells vs. bulk cancers remains unknown. Here, we show that stem cells derived from gliomas, GSCs, are up to 1,000-fold more sensitive to PIs (IC