EMAIL THIS PAGE TO A FRIEND
Haemophilia : the official journal of the World Federation of Hemophilia
Kinetic analysis and binding studies of a new recombinant human factor VIIa for treatment of haemophilia.
PMID 27995727
Abstract
LR769 is a new second-generation recombinant human Factor VIIa (rhFVIIa) developed for haemophilia treatment. We determined enzymatic properties of LR769 and its interaction with antithrombin, tissue factor, platelets and endothelial protein C receptor (EPCR), compared with NovoSevenRT. Kinetic enzyme assays and active site titration were used for enzymatic studies. Surface Plasmon Resonance (SPR) was used for determination of binding constants. Cellular binding was determined for platelets and cultured human umbilical vein endothelial cells (HUVEC). The dissociation constant (K Overall, LR769 exhibited characteristics similar to NovoSevenRT, but bound EPCR on HUVEC with somewhat higher affinity than NovoSevenRT.