EMAIL THIS PAGE TO A FRIEND
British journal of haematology
Phase 2 study of tabalumab, a human anti-B-cell activating factor antibody, with bortezomib and dexamethasone in patients with previously treated multiple myeloma.
PMID 28005265
Abstract
In this double-blind, Phase 2 study, 220 patients with relapsed/refractory multiple myeloma were randomly assigned 1:1:1 to receive placebo (N = 72), tabalumab 100 mg (N = 74), or tabalumab 300 mg (N = 74), each in combination with dexamethasone 20 mg and subcutaneous bortezomib 1·3 mg/m
Related Materials