EMAIL THIS PAGE TO A FRIEND
Journal of inorganic biochemistry
Potent anticancer activity of a new bismuth (III) complex against human lung cancer cells.
PMID 28006662
Abstract
The aim of this work is experimental study of an interesting bismuth(III) complex derived from pentadentate 2,6-pyridinedicarboxaldehyde bis(
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
17236
3-(Benzyldimethylammonio)propanesulfonate, BioXtra, ≥99.0% (HPCE)
C12H19NO3S