EMAIL THIS PAGE TO A FRIEND
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V
In vivo biodistribution of antihyperglycemic biopolymer-based nanoparticles for the treatment of type 1 and type 2 diabetes.
PMID 28007370
Abstract
This study aimed to assess the biodistribution of antihyperglycemic insulin-loaded alginate/dextran sulfate-based nanoparticles dual coated with chitosan and technetium-99m-albumin (
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S5502
Succinic dihydrazide, 96%
C4H10N4O2