EMAIL THIS PAGE TO A FRIEND
Neuroscience letters
Synergistic effect of olfactory ensheathing cells and alpha-crystallin on restoration of adult rat optic nerve injury.
PMID 28007643
Abstract
Olfactory enshesathing cells (OECs) and α-crystallin all can promote axonal regeneration after optic nerve injury. However, their mechanisms were different. Here, we study the synergistic effect of OECs and α-crystallin on the optic nerve regeneration. α-Crystallin was injected into vitreous cavity, and OECs were transplanted to the optic nerve injury area. The regeneration length of optic nerve were measured by anterograde tracing using cholera toxin subunit B (CTB). The survival of RGCs were assessed by counting the numbers of βIII-tubulin-labeled RGCs in a retinal whole mount. The results that OECs and α-crystallin all could promoted RGCs survival and axonal regeneration (P<0.01). Especially in the combination group, the length of axonal regeneration was 5.6mm after optic nerve injury for 3 months. These findings indicated that compared to OECs and α-crystallin alone, the combination of OECs and α-crystallin could promote axonal regeneration more effectively.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C6499 CTB, ≥98% (HPLC)
C16H13ClF3NO2