EMAIL THIS PAGE TO A FRIEND
Nanotechnology
Controlled synthesis of metallic iron nanoparticles and their magnetic hyperthermia performance in polyaniline composite nanofibers.
PMID 28008893
Abstract
Electrospun magnetic iron/polyaniline nanofibers with applicable heating performance in an AC magnetic field were developed. A new and low-cost method was introduced to synthesize metallic iron (Fe
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
205885
Palladium(II) chloride, ReagentPlus®, 99%
Cl2Pd