EMAIL THIS PAGE TO A FRIEND
Biochimica et biophysica acta
Alkyl-substituted phenylamino derivatives of 7-nitrobenz-2-oxa-1,3-diazole as uncouplers of oxidative phosphorylation and antibacterial agents: involvement of membrane proteins in the uncoupling action.
PMID 28011176
Abstract
In search for new effective uncouplers of oxidative phosphorylation, we studied 4-aryl amino derivatives of a fluorescent group 7-nitrobenz-2-oxa-1,3-diazol (NBD). In our recent work (Denisov et al., Bioelectrochemistry, 2014), NBD-conjugated alkyl amines (NBD-C
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
43608
3,3′-Dipropylthiadicarbocyanine iodide, suitable for fluorescence, ≥98.0% (TLC)
C25H27IN2S2