EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry
Small molecule mimics of DFTamP1, a database designed anti-Staphylococcal peptide.
PMID 28011203
Abstract
Antimicrobial peptides (AMPs) are important templates for developing new antimicrobial agents. Previously, we developed a database filtering technology that enabled us to design a potent anti-Staphylococcal peptide DFTamP1. Using this same design approach, we now report the discovery of a new class of bis-indole diimidazolines as AMP small molecule mimics. The best compound killed multiple S. aureus clinical strains in both planktonic and biofilm forms. The compound appeared to target bacterial membranes with antimicrobial activity and membrane permeation ability similar to daptomycin.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D2446
Daptomycin, cyclic lipopeptide antibiotic
C72H101N17O26