EMAIL THIS PAGE TO A FRIEND
International journal of pharmaceutics
Design and characterization of a magnetite/PEI multifunctional nanohybrid as non-viral vector and cell isolation system.
PMID 28011343
Abstract
It is described the reproducible formulation and complete physicochemical characterization of nanohybrids based on magnetite (Fe