EMAIL THIS PAGE TO A FRIEND
European journal of medicinal chemistry
Green synthesis and anticancer potential of chalcone linked-1,2,3-triazoles.
PMID 28011424
Abstract
A series of chalcone linked-1,2,3-triazoles was synthesized via cellulose supported copper nanoparticle catalyzed click reaction in water. The structures of all the compounds were analyzed by IR, NMR and Mass spectral techniques. All the synthesized products were subjected to 3-(4,5-dimethylthiazole-2-yl)-2,5-diphenyltetrazolium bromide (MTT) cytotoxicity assay against a panel of four human cancer cell lines (MCF-7, MIA-Pa-Ca-2, A549, HepG2) to check their anticancer potential. Compound 6h was found to be most active against all the tested cancer cell lines with IC
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D1054
Dihydrorhodamine 123, ≥95%
C21H18N2O3