EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Loss of Functionally Redundant p38 Isoforms in T Cells Enhances Regulatory T Cell Induction.
PMID 28011639
Abstract
The evolutionarily conserved protein kinase p38 mediates innate resistance to environmental stress and microbial infection. Four p38 isoforms exist in mammals and may have been co-opted for new roles in adaptive immunity. Murine T cells deficient in p38α, the ubiquitously expressed p38 isoform, showed no readily apparent cell-autonomous defects while expressing elevated amounts of another isoform, p38β. Mice with T cells simultaneously lacking p38α and p38β displayed lymphoid atrophy and elevated Foxp3