Fish physiology and biochemistry
Role of cyclooxygenase-mediated metabolites in lipid metabolism and expression of some immune-related genes in juvenile grass carp (Ctenopharyngodon idellus) fed arachidonic acid.
PMID 28012026
Cyclooxygenase (COX) catalyzes the conversion of arachidonic acid (ARA) to prostaglandins, and COX-mediated metabolites play important roles in the regulation of lipid metabolism and immunity in mammals. However, such roles of COX in fish remain largely unknown. In this study, we designed three semi-purified diets, namely ARA-free (control), ARA, and ARA + acetylsalicylic acid (ASA; a COX inhibitor), and used them to feed grass carp (27.65 ± 3.05 g) for 8 weeks. The results showed that dietary ARA significantly increased the amount of ARA in the hepatopancreas, muscle, and kidney (P < 0.05), whereas this increase was reduced by dietary ASA. The hepatopancreatic prostaglandin E2 content increased in the ARA group, and this increase was inhibited by ASA (P < 0.05). ARA decreased the lipid content in the hepatopancreas, whereas ASA recovered lipid content to a significant level (P < 0.05). ARA significantly decreased the messenger RNA (mRNA) expression levels of fatty acid synthase and stearoyl-CoA desaturase in the hepatopancreas (P < 0.05). However, ASA did not rescue the mRNA expression of these genes (P > 0.05). Interestingly, ARA significantly enhanced the level of peroxisome proliferator-activated receptor α gene expression, and this increase was attenuated by ASA (P < 0.05). Finally, ARA significantly enhanced the mRNA expression of myeloid differentiation factor 88 (MyD88) in the kidney, and ASA attenuated the expression of toll-like receptor 22 and MyD88 (P < 0.05). In conclusion, our findings suggest that COX metabolites play important roles in the inhibition of lipid accumulation in the hepatopancreas of grass carp fed with ARA and that regulation of gene expression promotes lipid catabolism rather than lipogenic activities. Additionally, these eicosanoids might participate in the upregulation of immunity-related genes in the kidney.
Related Materials
Product #
Molecular Formula
Add to Cart
47015-U PUFA No.2 (Animal Source), analytical standard
Pyridoxine, ≥98%