EMAIL THIS PAGE TO A FRIEND
Environmental pollution (Barking, Essex : 1987)
Perfluoroalkyl acids and their precursors in indoor air sampled in children's bedrooms.
PMID 28012670
Abstract
The contamination levels and patterns of perfluoroalkyl acids (PFAAs) and their precursors in indoor air of children's bedrooms in Finland, Northern Europe, were investigated. Our study is among the most comprehensive indoor air monitoring studies (n = 57) and to our knowledge the first one to analyse air in children's bedrooms for PFASs (17 PFAAs and 9 precursors, including two acrylates, 6:2 FTAC and 6:2 FTMAC). The most frequently detected compound was 8:2 fluorotelomer alcohol (8:2 FTOH) with the highest median concentration (3570 pg/m