EMAIL THIS PAGE TO A FRIEND
International journal of biological macromolecules
An efficient removal of RB5 from aqueous solution by adsorption onto nano-ZnO/Chitosan composite beads.
PMID 28013011
Abstract
In this study, the removal of Reactive Black 5 (RB-5) by nano-ZnO/Chitosan composite beads (nano-ZnO/CT-CB) from aqueous solution was investigated. ZnO nanoparticles were prepared by the via the microwave-assisted combustion technique. And then nano-ZnO/Chitosan composite beads were prepared by polymerization in the presence of nano-ZnO and chitosan. Characterization of composite beads were conducted using SEM, TEM, FTIR, TGA and XRD. Several important parameters influencing the removal of RB 5 such as contact time, pH and temperature were investigated systematically by batch experiments. At optimum conditions of pH 4 and adsorbent concentration of 0.2g, dye removal efficiency was found 76%. Langmuir, Freundlich and Temkin adsorption models were used to describe adsorption isotherms and constants. The maximum adsorption capacity (q