EMAIL THIS PAGE TO A FRIEND
Journal of molecular biology
H3K4 Methyltransferase Activity Is Required for MLL4 Protein Stability.
PMID 28013028
Abstract
Transcriptional enhancers play a key role in cell type-specific gene expression and cell fate transition. Enhancers are marked by histone H3K4 mono- and di-methylation (H3K4me1/2). The tumor suppressor MLL4 (KMT2D) is a major enhancer H3K4 mono- and di-methyltransferase with a partial functional redundancy with MLL3 (KMT2C). However, the functional role of MLL4 enzymatic activity remains elusive. To address this issue, we have generated MLL4 enzyme-dead knock-in (KI) embryonic stem (ES) cells and mice, which carry Y5477A/Y5523A/Y5563A mutations in the enzymatic SET domain of the MLL4 protein. Homozygous MLL4 enzyme-dead KI (Mll4
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M7449 MG-132, Ready Made Solution, ≥90% (HPLC)