EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Iron (II, III) oxide/multi-walled carbon nanotube composite as solid-phase extraction sorbent followed by ultra-high performance liquid chromatography tandem mass spectrometry for simultaneous determination of zearalenone and type A trichothecenes in Salviae miltiorrhizae Radix et Rhizoma (Danshen).
PMID 28017565
Abstract
For the first time, a reliable solid-phase extraction (SPE) procedure using iron (II, III) oxide (Fe