EMAIL THIS PAGE TO A FRIEND
Nitric oxide : biology and chemistry
Nitrite-derived nitric oxide reduces hypoxia-inducible factor 1α-mediated extracellular vesicle production by endothelial cells.
PMID 28017872
Abstract
Extracellular vesicles (EVs) are small, spherical particles enclosed by a phospholipid bilayer (∼30-1000 nm) released from multiple cell types, and have been shown to have pathophysiological roles in a plethora of disease states. The transcription factor hypoxia-inducible factor-1 (HIF-1) allows for adaptation of cellular physiology in hypoxia and may permit the enhanced release of EVs under such conditions. Nitric oxide (NO) plays a pivotal role in vascular homeostasis, and can modulate the cellular response to hypoxia by preventing HIF-1 accumulation. We aimed to selectively target HIF-1 via sodium nitrite (NaNO Endothelial (HECV) cells were exposed to hypoxic conditions (1% O Endothelial (HECV) cells exposed to hypoxia (1% O These data provide evidence that hypoxia enhances the release of EVs in endothelial cells, and that this is mediated by HIF-1α, but not HIF-2α. Furthermore, the reduction of NO