EMAIL THIS PAGE TO A FRIEND
Digestive diseases and sciences
Soluble Syndecan-1: A Novel Biomarker of Small Bowel Mucosal Damage in Children with Celiac Disease.
PMID 28025744
Abstract
Syndecan-1 (SDC1) is essential for maintaining normal epithelial barrier. Shedding of SDC1 ectodomain, reflected by serum soluble syndecan-1 (SSDC1) levels, is regulated by inflammation. Increased intestinal permeability plays a central role in celiac disease (CD). The association between SSDC1 levels and mucosal damage in CD has not been evaluated. To evaluate serum SSDC1 levels in children with CD and to determine its relationship with histological grading classified by modified Marsh criteria. This is a cross-sectional, pilot study, in which serum SSDC1 was analyzed by ELISA in a cohort of 49 untreated children with CD and 15 children with nonspecific abdominal pain (AP). CD was diagnosed based on positive celiac serology and small intestinal biopsy. SSDC1 levels at the time of biopsy were correlated with Marsh grading. Controls were defined by AP, negative celiac serology, normal upper endoscopy, and small intestinal biopsies. SSDC1 levels were significantly higher in CD patients compared to AP controls (116.2 ± 161 vs. 41.3 ± 17.5 ng/ml, respectively, p < 0.01). SSDC1 levels were significantly higher in patients with Marsh 3c lesion compared to AP controls (170.6 ± 201 vs. 41.3 ± 17.5 ng/ml, respectively, p < 0.05). SSDC1 concentrations displayed a significant correlation with mucosal damage defined by Marsh (r = 0.39, p < 0.05). This is the first study demonstrating elevated levels of serum SSDC1 in children with CD. Our results suggest that SSDC1 is a potentially novel marker of intestinal mucosal damage in patients with CD. Its applicability as a surrogate biomarker in CD remains to be determined.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
RAB0736 Human SDC1 / Syndecan-1 ELISA Kit