EMAIL THIS PAGE TO A FRIEND
Differentiation; research in biological diversity
Disruption of homeobox containing gene, hbx9 results in the deregulation of prestalk cell patterning in Dictyostelium discoideum.
PMID 28027501
Abstract
We have earlier classified the homeobox containing genes from Dictyostelium discoideum and identified 5 genes belonging to the TALE class. Here, we have characterized hbx9, a member of TALE class. In situ hybridization results show it to be preferentially expressed in the prestalkA (pstA) cells but not in the prestalkO (pstO) cells. Disruption of the hbx9 gene in Ax2 cells delay initiation of development and also slows cell proliferation. There was decreased cadA expression during the development of hbx9
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
199524
Chlorophenol Red, indicator grade
C19H12Cl2O5S