EMAIL THIS PAGE TO A FRIEND
Chemosphere
The SSRI fluoxetine exhibits mild effects on the reproductive axis in the cichlid fish Cichlasoma dimerus (Teleostei, Cichliformes).
PMID 28030789
Abstract
Among the wide variety of pharmaceuticals released into the environment, Fluoxetine (FLX), a selective serotonin reuptake inhibitor, is one of the most prescribed for the treatment of major depression. It inhibits serotonin (5-HT) reuptake at the presinaptic membrane, increasing serotonergic activity. In vertebrates, including fish, the serotonergic system is closely related to the Hypothalamic Pituitary Gonadal (HPG) axis which regulates reproduction. As FLX can act as an endocrine disrupting compound (EDC) by affecting several reproductive parameters in fish, the aim of this study was to provide an integral assessment of the potential effect of FLX on the reproductive axis of the Neotropical freshwater fish Cichlasoma dimerus. Adult fish were intraperitoneally injected with 2 μg g