EMAIL THIS PAGE TO A FRIEND
Behavioral and neural biology
Perinatal bromopride treatment: effects on sexual behavior of male and female rats.
PMID 2803170
Abstract
Effects of different perinatal bromopride treatments on sexual behavior were examined in adult male and female rats of Wistar origin. Female rats of mothers treated with bromopride (BRO) during lactation (VB group) and during pregnancy and lactation (BB group) showed lower lordosis quotients than those of controls (VV group). Females of mothers treated with BRO only during pregnancy (BV group) showed no differences in lordosis quotient when compared to the VV group. There were no significant differences in male sexual behavior between experimental and control groups. The effect of bromopride on the control of sexual behavior is discussed.